Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wire Gauzes, Plain

Wire Gauzes, Plain

Wire Gauze, Plain, 6" x 6", 15×15 cm (6×6")
Catalog # 470188-998
Supplier #:  WGPL66
Wire Gauzes, Plain
Catalog # 470188-998
CAS Number:  
Wire Gauzes, Plain
Wire Gauzes, Plain

Wire Gauze, Plain, 6" x 6", 15×15 cm (6×6")

CAS Number:  
Clearance
$41.20
Log in for availability
Each (1UN) Paquete de 10 (1UN)
Product Details & Documents

Sturdy wire mesh for supporting glassware over a Bunsen burner.

  • Long lasting, heat- and tear-resistant iron

  • Available in a variety of sizes

Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.

Specifications

Specifications

  • Description:
    Wire Gauze, Plain, 6" x 6"
  • Size:
    15×15 cm (6×6")

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.