Order Entry
Mexico
Orders LinkContactUsLinkComponent
Banana Plug Patch Cords

Banana Plug Patch Cords

Patch cord, black, 15 cm, banana plugs both ends
Catalog # 470188-740
Supplier #:  1350302190001
Banana Plug Patch Cords
Catalog # 470188-740
Banana Plug Patch Cords
Banana Plug Patch Cords

Patch cord, black, 15 cm, banana plugs both ends

Price
$52.56
Log in for availability
Each (1UN) Paquete de 6 (1UN)
Product Details & Documents

Double ended banana plug jumper wires.

  • Available in red or black
  • 15 cm (6") or 61 cm (24") long

Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.

Specifications
  • Description:
    Patch cord, black, 15 cm, banana plugs both ends
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.