Avian Development Slides
Chick, 18-Hour, Whole mount, Schneider's
470182-442
Explore Developmental Biology.
- Investigate varying ages of developing chicks
- Illustrates specific structures at developmental stages
- Clearly distinguishable features
Chickens are historic model organisms for developmental studies. They are particularly useful in comparative anatomy and vertebrate embryology studies because of the similarities in embryological development across many vertebrate species, including humans.
: Choose from a variety of individual slides ranging from 12-92 hr. development in an assortment of slide types.
- Description:Chick, 18-Hour
- Stain:Schneider's
- Orientation:Whole mount