Ward's® Lichen Slides
Foliose, Section, Quadruple
Catalog # 470181-104
Supplier: Avantor
Jump to:
Product Details & Documents
A composite of fungi and algae.
Ward's produced slides
- Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.
Specifications
- Description:Foliose
- Stain:Quadruple
- Orientation:Section