Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polystyrene Storage Jars

Polystyrene Storage Jars

4 oz., 58 mm
Catalog # 470178-182
Supplier #:  181633
Polystyrene Storage Jars
Catalog # 470178-182
Polystyrene Storage Jars
Polystyrene Storage Jars

4 oz., 58 mm

Price
$13.43
Log in for availability
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents

Ideal for specimen storage.

  • Clear polystyrene
  • Wide-mouth
  • Aligned with GLOBE protocols

Ordering information: Metal jar caps sold separately.

Specifications
  • Cap Size:
    58 mm
  • Capacity:
    4 oz.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.