Simple Cuboidal Epithelium Slides
Simple Cuboidal Epithelium, Cross section, Quadruple
470177-562
: Avantor
Aid in secretion and absorption of organs.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Choose from several available stains and preparations for these tissue sections to get the view you're looking for. Slides are made from mammal kidney, and demonstrate cuboidal cells in tubules. Thin section slide is plastic-embedded for greater viewing clarity.
- Description:Simple Cuboidal Epithelium
- Stain:Quadruple
- Orientation:Cross section