Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Wire Gauzes, Plain

Wire Gauze, Plain, 4" x 4", 10×10 cm (4×4")
Catalog # 470176-062
 :  WGPL44
Wire Gauzes, Plain
Catalog # 470176-062
CAS Number:  
Wire Gauzes, Plain
Wire Gauzes, Plain

Wire Gauze, Plain, 4" x 4", 10×10 cm (4×4")

CAS Number:  
Price
$20.06
Log in for availability
Each (1UN) Paquete de 10 (1UN)
Product Details & Documents

Sturdy wire mesh for supporting glassware over a Bunsen burner.

  • Long lasting, heat- and tear-resistant iron

  • Available in a variety of sizes

Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.

Specifications

Specifications

  • Description:
    Wire Gauze, Plain, 4" x 4"
  • Size:
    10×10 cm (4×4")

Specifications