Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polypropylene Cylinders

Polypropylene Cylinders

250 ml, 40.60 mm, 2.0 ml
Catalog # 470177-818
Supplier:   VWR International
Polypropylene Cylinders
Catalog # 470177-818
Supplier:  VWR International
Polypropylene Cylinders
Polypropylene Cylinders

250 ml, 40.60 mm, 2.0 ml

Price
$38.70
Log in for availability
Each (1UN) Case de 6 (1UN)
Product Details & Documents

Unbreakable Polypropylene.

  • Permanent, molded, single scale metric markings
  • Flared top rims and large spouts facilitate pouring
  • Hexagonal shaped bases prevent tipping and rolling
  • Autoclavable at 121°C

Chemically resistant to most acids, bases, alcohols, esters, and ketones.

Documents

Specifications
  • Ext.Ø:
    40.60 mm
  • Graduation Interval:
    2.0 ml
  • Capacity:
    250 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.