Order Entry
Mexico
Orders LinkContactUsLinkComponent
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Compound Coarse and Fine Focus Beginner Microscope, Corded Tungsten
Catalog # 470177-094
Supplier #:  5585000
Boreal Science Compound Beginner Microscopes
Catalog # 470177-094
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Compound Coarse and Fine Focus Beginner Microscope, Corded Tungsten
Price
$3,304.64
Log in for availability
Each (1UN)
Product Details & Documents
Boreal beginner microscopes are ideal for introducing students to microscopy.
  • All-Glass-Coated Optics
  • All-Metal Construction
  • Parfocal and Parcentered
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It's scaled down for younger students and is simple to use, but it also prepares students for the transition to high school instruction.
Rugged metal components are engineered for years of reliable performance. Student-proof features minimize loss of parts.
 : This product includes a manufacturer's warranty.
Specifications
  • Description:
    Compound Coarse and Fine Focus Beginner Microscope
  • Illumination:
    Corded Tungsten