Ward's® Buttercup (Ranunculus) Slides
Young and mature root, Cross section, Quadruple
Catalog # 470176-880
Supplier: Avantor
Jump to:
Product Details & Documents
A herbaceous dicot.
Ward's produced slides
- Rigorous quality control standards
Compare the anatomy of a common dicot. Ranunculus is most often used to show root development. The large cortex and central vascular cylinder make tissue identification easy.
Specifications
- Description:Young and mature root
- Stain:Quadruple
- Orientation:Cross section