Ward's® Pine Structure Microscope Slides
Pinus resinosa leaf; 2 needle, Cross section, Quadruple stained
Catalog # 470176-872
Supplier: Avantor
Jump to:
Product Details & Documents
Anatomy of a gymnosperm.
- All aspects of pine live history represented
- Prepared by expert technicians
Explore all aspects of the gymnosperm life cycle. Pinus is a common educational representative of the gymnosperms exhibiting the general characteristics of the clade. Root, stem, leaf, and reproductive structures are all available.
Specifications
- Description:Pinus resinosa leaf; 2 needle
- Stain:Quadruple stained
- Orientation:Cross section