Tri Pour Beakers
50 ml, 100 ml, 250 ml, 400 ml, 800 ml, 1000 ml, Tri-Pour Beakers, Set of 6 different sizes
Catalog # 470174-590
Supplier: DYN-A-MED PRODUCTS
Supplier #: 80091
Jump to:
Product Details & Documents
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
Specifications
- Capacity:50 ml, 100 ml, 250 ml, 400 ml, 800 ml, 1000 ml
- Description:Tri-Pour Beakers, Set of 6 different sizes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



