Polyethylene Disposable Gloves
Polyethylene gloves, L, Box/100
Catalog # 470153-724
Supplier: 3G PACKAGING, INC.
Supplier #: GDPE-LG
Jump to:
Product Details & Documents
Ultra sensitive.
- Ambidextrous
- Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Specifications
- Glove size:L
- Description:Polyethylene gloves
- Packaging:Box/100
- Cat. No.:470153-724
- Disposable / reusable:Disposable
- Powdered / powder-free:Powder-free
- Ambidextrous / hand-specific:Ambidextrous