Order Entry
Mexico
Orders LinkContactUsLinkComponent
Precision pH Test Strips
Precision pH Test Strips
pH paper strips, 10.0–13.0
Catalog # 470150-880
Supplier #:  PH1013-1B-50
Precision pH Test Strips
Catalog # 470150-880
Precision pH Test Strips
Precision pH Test Strips
pH paper strips, 10.0–13.0
Price
$167.38
Log in for availability
Each (1UN)
Product Details & Documents
The indicator on these strips won’t contaminate the solutions the strips are testing.
  • Vinyl-backed
  • Non-contaminating indicator
  • Fifty strips per package
Durable pH testing strips that are backed with vinyl.
Specifications
  • Description:
    pH paper strips
  • pH Range:
    10.0–13.0