Acrylic Buret
50 ml, 0.1 ml
Catalog # 470148-702
Supplier: VWR International
Jump to:
Product Details & Documents
These hardy burets are ideal for classroom use.
- Acrylic
- Transparent
- Leakproof Stopcock Included
- Burets Unaffected by Most Dilute Aqueous Solutions
- Heavy-Duty
The transparent, acrylic burets are supplied with self-lubricating, leakproof PMP stopcock.
They are unaffected by most dilute aqueous solutions except ammonia and HF and are not recommended for use with ethanol or organic solvents.
Specifications
- Graduation Interval:0.1 ml
- Capacity:50 ml



