Refraction Blocks
Refraction Block, 75 x 50 x 15 mm
470105-730
Supplier #: RCB075
Rectangular blocks in a variety of sizes and are designed for use as an index of refraction plate.
- Quality Lucite block
- Optically clear sides
This Lucite block allows you to investigate refraction and internal reflection.
- Description:Refraction Block
- Dimensions:75 x 50 x 15 mm