Order Entry
Mexico
Orders LinkContactUsLinkComponent
Friction Rods
Friction Rods
Hard Nylon, Flat Ended
Catalog # 470104-254
Supplier:   CURBELL PLASTICS
Supplier #:  BLNYLROD
Friction Rods
Catalog # 470104-254
Supplier:  CURBELL PLASTICS
Friction Rods
Friction Rods
Hard Nylon, Flat Ended
Price
$99.15
Log in for availability
Each (1UN)
Product Details & Documents
For your Electrostatic Needs!
  • Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Specifications
  • Description:
    Hard Nylon, Flat Ended