Luer Lock Stop Cocks
Stopcock, 2-Way, Luer Lock, Male/ Female
Catalog # 470102-722
Supplier: S17 SCIENCE LLC.
Supplier #: EQ 066
Jump to:
Product Details & Documents
Assortment of 2-way stopcocks, syringes, and syringe extenders control the flow of liquids and gases, and make gas-tight connections.
- Polypropylene
Specifications
- Description:Stopcock, 2-Way, Luer Lock, Male/ Female