Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Bottle, Dispensing, 30 mL
Catalog # 470100-718
Supplier #:  300 & 351
Polyethylene Dispensing Bottles
Catalog # 470100-718
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Bottle, Dispensing, 30 mL
Price
$256.91
Log in for availability
Paquete de 12
Product Details & Documents
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Specifications
  • Description:
    Bottle, Dispensing, 30 mL