Coplin Staining Jars
Up to 10 Slides, Coplin Staining Jar, Glass
Catalog # 470175-194
Supplier: VARIETY GLASS INC
Supplier #: 674
Jump to:
Product Details & Documents
Coplin Jar with Screw-on Top Prevents Spills.
- Quality slide making supplies
- Slide staining tools
The screw-on cover prevents evaporation and contamination when staining. Each jar holds five single slides or ten back to back.
Specifications
- Capacity:Up to 10 Slides
- Description:Coplin Staining Jar, Glass