Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polymethylpentene Cylinders

Polymethylpentene Cylinders

1000 ml, 10.0 ml
Catalog # 470174-186
Supplier:   DYNALON
Supplier #:  237895
Polymethylpentene Cylinders
Catalog # 470174-186
Supplier:  DYNALON
Polymethylpentene Cylinders
Polymethylpentene Cylinders

1000 ml, 10.0 ml

Price
$759.95
Log in for availability
Each (1UN) Case de 4 (1UN)
Product Details & Documents

These polymethylpentene cylinders provide glass-like clarity and can withstand temperatures up to 350°F.

  • Heat-resistant
  • Shatterproof

The cylinders may be autoclaved, and they are chemically resistant to most acids, bases, alcohols, and aldehydes. All cylinders are calibrated to contain.

Specifications
  • Graduation Interval:
    10.0 ml
  • Capacity:
    1000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.