Order Entry
Mexico
Orders LinkContactUsLinkComponent
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, heavy metals, 50 tests
Catalog # 470173-838
Supplier #:  480309
Single Factor Water Test Kits
Catalog # 470173-838
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, heavy metals, 50 tests
Price
$577.77
Log in for availability
Each (1UN) Case de 12 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Specifications
  • Description:
    Single factor water test kit, heavy metals, 50 tests