$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
PH PAPER INSTA CHECK 0-13 PH 5/32 X 15FT
470153-246
:
MICRO ESSENTIAL LABORATORY I
PH PAPER INSTA CHECK 0-13 PH 5/32 X 15FT
470153-246
:
MICRO ESSENTIAL LABORATORY I
:
M1313
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International