Order Entry
Mexico
Orders LinkContactUsLinkComponent
Waterproof Field Notebooks
Waterproof Field Notebooks
Waterproof Field Notebooks, Field; 32 Sheets, 4⅝", 7"
Catalog # 470152-782
Supplier:   J.L. DARLING LLC.
Supplier #:  353
Waterproof Field Notebooks
Catalog # 470152-782
Supplier:  J.L. DARLING LLC.
Waterproof Field Notebooks
Waterproof Field Notebooks
Waterproof Field Notebooks, Field; 32 Sheets, 4⅝", 7"
Price
$347.73
Log in for availability
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents
An absolute must when conducting field studies.
  • Take legible notes in any conditions
  • Easy to spot if accidentally dropped
  • Aligned with GLOBE protocols
Unique all–weather paper lets you take and keep legible notes regardless of the conditions, and the bright yellow cover is easy to spot if accidentally dropped. The field version has one page for data values and on the facing page it has a grid for drawing.
Specifications
  • Description:
    Waterproof Field Notebooks, Field; 32 Sheets
  • Length:
    7"
  • Width:
    4⅝"