Order Entry
Mexico
Orders LinkContactUsLinkComponent
Cylinder Guards

Cylinder Guards

Cylinder Guards, For Cylinder Cap., 25 – 50 mL
Catalog # 470151-206
Supplier #:  VCYCG2
Cylinder Guards
Catalog # 470151-206
Cylinder Guards
Cylinder Guards

Cylinder Guards, For Cylinder Cap., 25 – 50 mL

Price
$58.05
Log in for availability
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents

Black sponge rubber guards prevent breakage and rolling.

  • Hexagonal
  • 13 mm thick
Specifications
  • Description:
    Cylinder Guards, For Cylinder Cap., 25 – 50 mL
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.