United Scientific™ Push/Pull Spring Scales
Push/pull spring scale, red, 2000 g/20 N, graduation interval: 0.4 N
Catalog # 470150-474
Supplier: UNITED SCIENTIFIC SUPPLIES
: PS2000
United Scientific™ Push/Pull Spring Scales
Catalog # 470150-474
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Product Details & Documents
Versatile spring scale.
- Weighs in Grams and Newtons
- Color coded to differentiate capacity
These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.
: Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.
: This product is for ages six and up.
Specifications
- Description:Push/pull spring scale, red
- Weighing Capacity:2000 g/20 N, graduation interval: 0.4 N



