Order Entry
Mexico
Orders LinkContactUsLinkComponent
Low-Form Evaporating Dish
Low-Form Evaporating Dish
Low-Form Evaporating Dish, Size 1, 30 mL
Catalog # 470149-550
Supplier #:  J-281
Low-Form Evaporating Dish
Catalog # 470149-550
Low-Form Evaporating Dish
Low-Form Evaporating Dish
Low-Form Evaporating Dish, Size 1, 30 mL
Price
$80.09
Log in for availability
Each (1UN) Case de 24 (1UN)
Product Details & Documents
High-Quality Labware, Low Prices
Our laboratory porcelain is high-quality, yet affordably priced. This low-form evaporating dish has a pourout. It is glazed entirely on the inside and over the upper section of the outside. Note: Fragile, use care when handling.
Specifications
  • Description:
    Low-Form Evaporating Dish, Size 1, 30 mL