Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Measuring Pipettes

25 ml, VWR® Measuring Pipette, 0.1 ml
  470149-276
VWR® Measuring Pipettes
  470149-276
VWR® Measuring Pipettes
VWR® Measuring Pipettes

25 ml, VWR® Measuring Pipette, 0.1 ml

 
$686.05
   
Paquete de 6 Case de 2 (6UN)
 

These high-quality borosilicate, Mohr-type pipettes meet ASTM standards.

  • Color-coded by size
  • Calibrated 'to deliver'
  • Measurements are accurate to ±0.01 ml

The pipettes feature colored graduations in 0.1 increments that are enameled into the glass to prevent fading.

 

 
  • Capacity:
    25 ml
  • Description:
    VWR® Measuring Pipette
  • Subdivisions:
    0.1 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.