Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Test Tubes

VWR® Test Tubes

27 ml, 0-1, 18×150 mm
  470149-264
VWR® Test Tubes
  470149-264
VWR® Test Tubes
VWR® Test Tubes

27 ml, 0-1, 18×150 mm

 
$838.43
   
Paquete de 72 Case de 10 (72UN)
 

VWR® Test tubes are made from high-quality glass.

  • Borosilicate glass construction
  • Uniform thickness

Each tube features a strong lip.

Certifications: ASTM standards.

Documents

 
  • Capacity:
    27 ml
  • Stopper Size:
    0-1
  • O.D.×L:
    18×150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.