Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Test Tubes

VWR® Test Tubes

34 ml, 2-3, 20×150 mm
Catalog # 470149-262
Supplier:   VWR International
VWR® Test Tubes
Catalog # 470149-262
Supplier:  VWR International
VWR® Test Tubes
VWR® Test Tubes

34 ml, 2-3, 20×150 mm

Price
$850.79
Log in for availability
Paquete de 72 Case de 10 (72UN)
Product Details & Documents

VWR® Test tubes are made from high-quality glass.

  • Borosilicate glass construction
  • Uniform thickness

Each tube features a strong lip.

 : ASTM standards.

Documents

Specifications
  • Capacity:
    34 ml
  • Stopper Size:
    2-3
  • O.D.×L:
    20×150 mm