Order Entry
Mexico
Orders LinkContactUsLinkComponent
Disposable Micropipet Tips
Disposable Micropipet Tips
200 - 1000 µl, Blue, 1000 Tips/Bag
Catalog # 470149-166
Supplier:   VWR International
Disposable Micropipet Tips
Catalog # 470149-166
Supplier:  VWR International
Disposable Micropipet Tips
Disposable Micropipet Tips
200 - 1000 µl, Blue, 1000 Tips/Bag
Price
$284.62
Log in for availability
Case (1,000UN)
Product Details & Documents
High quality micropipette tips to meet any lab's needs.
  • Certified DNase, RNase and pyrogen free
  • Material: PP
  • Autoclavable
  • Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Specifications
  • Volume:
    200 - 1000 µl
  • Tip Color:
    Blue
  • Packed:
    1000 Tips/Bag