Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Linear Spring Scales
VWR® Linear Spring Scales
5000 g x 100 g (50 N x 5 N)
Catalog # 470149-050
VWR® Linear Spring Scales
Catalog # 470149-050
Supplier:  
VWR® Linear Spring Scales
VWR® Linear Spring Scales
5000 g x 100 g (50 N x 5 N)
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SPRING SCALE 5000GX100G / 50NX1N VWR Catalog #470201-024
Product 470201-024 is a replacement for 470149-050 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Designed for Heavy Duty Student Use
  • Feature easy to read markings
  • Durable for years of regular student use
  • Length: 14 cm
  • Each scale is easily zeroed
Each has a plated steel suspension ring at the top, and a hook at the bottom.
Specifications
  • Weighing Capacity:
    5000 g x 100 g (50 N x 5 N)