Illustrated Dissection Guides
Mammalian Eye Guide, 6, 22 cm, 14 cm
470146-788
: 420.4204.1
These economical and concise guides contain the right amount of information for reference during dissection.
- Basic Facts on Specimens
- Convenient Size
- Paperback
- Color Illustrations
- Step-by-Step Instructions
Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.
The paperback guides are small, so students can have them whenever they're needed.
- Description:Mammalian Eye Guide
- Length:22 cm
- No. of Pages:6
- Width:14 cm



