Order Entry
Mexico
Orders LinkContactUsLinkComponent
Illustrated Dissection Guides

Illustrated Dissection Guides

Mammalian Kidney Guide, 8, 22 cm, 14 cm
Catalog # 470146-776
Supplier:   RANACO CORP SE
Supplier #:  420.4194.1
Illustrated Dissection Guides
Catalog # 470146-776
Supplier:  RANACO CORP SE
Illustrated Dissection Guides
Illustrated Dissection Guides

Mammalian Kidney Guide, 8, 22 cm, 14 cm

Price
$186.54
Log in for availability
Each (1UN)
Product Details & Documents

These economical and concise guides contain the right amount of information for reference during dissection.

  • Basic Facts on Specimens
  • Convenient Size
  • Paperback
  • Color Illustrations
  • Step-by-Step Instructions

Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.

The paperback guides are small, so students can have them whenever they're needed.

Specifications
  • Description:
    Mammalian Kidney Guide
  • Length:
    22 cm
  • No. of Pages:
    8
  • Width:
    14 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.