Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Illustrated Dissection Guides

Earthworm Guide, 8, 22 cm, 14 cm
Catalog # 470146-770
Supplier:   RANACO CORP SE
 :  420.4014.1
Illustrated Dissection Guides
Catalog # 470146-770
Supplier:  RANACO CORP SE
Illustrated Dissection Guides
Illustrated Dissection Guides

Earthworm Guide, 8, 22 cm, 14 cm

Price
$218.00
Log in for availability
Each (1UN)
Product Details & Documents

These economical and concise guides contain the right amount of information for reference during dissection.

  • Basic Facts on Specimens
  • Convenient Size
  • Paperback
  • Color Illustrations
  • Step-by-Step Instructions

Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.

The paperback guides are small, so students can have them whenever they're needed.

Specifications
  • Description:
    Earthworm Guide
  • Length:
    22 cm
  • No. of Pages:
    8
  • Width:
    14 cm