Order Entry
Mexico
Orders LinkContactUsLinkComponent
Illustrated Dissection Guides

Illustrated Dissection Guides

Turtle Guide, 56, 22 cm, 14 cm
  470146-762
Supplier #:  420.4114.1
Illustrated Dissection Guides
  470146-762
Illustrated Dissection Guides
Illustrated Dissection Guides

Turtle Guide, 56, 22 cm, 14 cm

 
$311.28
   
Each (1UN)
 

These economical and concise guides contain the right amount of information for reference during dissection.

  • Basic Facts on Specimens
  • Convenient Size
  • Paperback
  • Color Illustrations
  • Step-by-Step Instructions

Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.

The paperback guides are small, so students can have them whenever they're needed.

 
  • Description:
    Turtle Guide
  • Length:
    22 cm
  • No. of Pages:
    56
  • Width:
    14 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.