Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Burets
PYREX® Burets
Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
Catalog # 470146-252
   Low Manufacturing Impact
Supplier:   CORNING CA
Supplier #:  2116-GTO2
PYREX® Burets
Catalog # 470146-252
Supplier:  CORNING CA
   Low Manufacturing Impact
PYREX® Burets
PYREX® Burets
Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
Price
$3,840.00
Log in for availability
Paquete de 12
Product Details & Documents
Educational grade Pyrex burets.
  • Heavy duty
  • Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
Documents
Specifications
  • Description:
    Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
  • Cat. No.:
    470146-252
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact