Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rectangular Glass Coverslips

Rectangular Glass Coverslips

Glass cover slips, rectangular, 40×22 mm, 0.2 mm, #2
Catalog # 470145-746
Número de proveedor #:  94-22402
Rectangular Glass Coverslips
Catalog # 470145-746
Rectangular Glass Coverslips
Rectangular Glass Coverslips

Glass cover slips, rectangular, 40×22 mm, 0.2 mm, #2

Price
$87.42
Log in for availability
Onza
Product Details & Documents

Traditional rectangular coverslip style

  • Quality slide making supplies
  • Traditional coverslip type

Traditional style coverslips to use when making slides in the laboratory.

#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.

Specifications
  • Numbering:
    #2
  • Description:
    Glass cover slips, rectangular
  • L×W:
    40×22 mm
  • Thickness:
    0.2 mm