Slab Easels
Slab Easels, Thick Slab Easel
Catalog # 470145-234
Supplier #: 35
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SLAB PLASTIC DISPLAY STAND PK/4 Catalog #470018-778
Product 470018-778 is a functional alternative to 470145-234 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Product Details & Documents
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
Specifications
- Description:Slab Easels, Thick Slab Easel