Order Entry
Mexico
Orders LinkContactUsLinkComponent
Curriculum Mastery Posters

Curriculum Mastery Posters

Photosynthesis And Respiration, Laminated, 89 cm, 58.5 cm
Catalog # 470144-522
Supplier #:  346105
Curriculum Mastery Posters
Catalog # 470144-522
Curriculum Mastery Posters
Curriculum Mastery Posters

Photosynthesis And Respiration, Laminated, 89 cm, 58.5 cm

Price
$563.25
Log in for availability
Each (1UN) Paquete de 20 (1UN)
Product Details & Documents

A Graphic Look At Key Science Concepts

  • Detailed illustrations
  • Representative photographs
  • Colorful depictions

Large, reference posters facilitate comprehension of key, standards-based concepts. Each poster is laminated for years of use.

Cells
Vivid images compare and contrast the similarities and differences between plant and animal cells, including detailed representation of each organelle and examples of various specialized cells.

Osmosis and Diffusion
Examine the structure of cell membranes, the processes of diffusion and osmosis and the differences between them.

Modern Genetics
Thorough, visual overview of the structure and function of chromosomes, genes and DNA also provides a step-by-step procedure for DNA fingerprinting.

Photosynthesis and Respiration
Take a comprehensive look at the steps of photosynthesis and the two stages of respiration.
Microlife
Illustrations and photographs show a typical bacterial cell, various fungi types, as well as amoeba, paramecium and euglena.

Specifications
  • Width:
    58.5 cm
  • Description:
    Photosynthesis And Respiration
  • Length:
    89 cm
  • Finish:
    Laminated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.