Parafilm
Parafilm, Roll Size, in. x ft.: 20 x 50
Catalog # 470144-226
Supplier: HEATHROW SCIENTIFIC SE
Supplier #: 120265
Jump to:
Product Details & Documents
Flexible, self-adhesive, waterproof plastic for closures, mounting and more.
Adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing petri dish edges. Excellent resistance to air, other gases, alcohol, common acids, and moderately concentrated alkalies.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Parafilm, Roll Size, in. x ft.: 20 x 50