Order Entry
Mexico
Orders LinkContactUsLinkComponent
Specific Gravity Set
Specific Gravity Set
Specific Gravity Set, Set of 4
  470137-628
Specific Gravity Set
  470137-628
Specific Gravity Set
Specific Gravity Set
Specific Gravity Set, Set of 4
$14.85
  31/12/2026
Paquete de 4
Teach About Gravity
  • Metals included are: aluminum, brass, copper and steel
  • Each cylinder is 5cm long and 13mm diameter
  • Purchase as a set or individually
Determine the gravity of common metals using this set of specific gravity cylinders. Perfect for specific gravity determinations in the classroom. 
Caution: WARNING: CHOKING HAZARD-Not for children under 3 years.
  • Description:
    Specific Gravity Set, Set of 4