Order Entry
Mexico
Orders LinkContactUsLinkComponent
Collision Track
Collision Track
Motorized Cart
Catalog # 470128-702
Supplier:   VWR International
Collision Track
Catalog # 470128-702
Supplier:  VWR International
Collision Track
Collision Track
Motorized Cart
Clearance
$26.65
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN)
Product Details & Documents
This track includes everything needed to demonstrate three basic concepts of physics: acceleration, collision, and force.
  • Instructions Provided
  • Two Carts Provided
  • Two Sets of Slotted Masses Provided
  • Two Photogate Support Rods Provided
  • Physics Topics Include Acceleration, Collision, and Force
Change the speed of the cart by using the steel slotted masses, or accelerate it by using the provided pulley to demonstrate the use of external forces.
The provided carts have spring bands on one side for elastic collisions and Velcro on the other for inelastic collisions.
Ordering information: This track includes a 1.2 m track, two carts, a pulley, two sets of slotted masses (10 g, 20 g, and 50 g), two photogate support rods, and instructions.
Specifications
  • Description:
    Motorized Cart