Squeeze-Jet Bottles
Squeeze-Jet Bottles, 60 ml
Catalog # 470123-922
Supplier: DYNALON
Supplier #: 524165-0060
Jump to:
Product Details & Documents
Clear Polyethylene.
- Use for drop dispensing or for a jet spray
- Air-tight seal is made when the dispenser nozzle is closed
Specifications
- Capacity:60 ml
- Description:Squeeze-Jet Bottles