Litmus Test Sheets
Litmus Test Sheets, Litmus Blue
Catalog # 470123-140
Supplier: PRECISION LABORATORIES SE
: 280-24-810
Jump to:
Product Details & Documents
These litmus test sheets are available in litmus red (ACS) or litmus blue (ACS).
- Individual Sheet Length: 8"
- Individual Width: 10"
- Twenty-Four Sheets in One Package
- Litmus Blue and Litmus Red Available
These individual sheets of litmus test papers are 8 x 10".
One package has 24 sheets.
: One package has 24 sheets.
Specifications
- Description:Litmus Test Sheets, Litmus Blue



