Order Entry
Mexico
Orders LinkContactUsLinkComponent
Stackable Banana Plug Connecting Wires

Stackable Banana Plug Connecting Wires

Red Stackable Banana Plug Connecting Wires, 36"
Catalog # 470122-134
Supplier:   VWR International
Stackable Banana Plug Connecting Wires
Catalog # 470122-134
Supplier:  VWR International
Stackable Banana Plug Connecting Wires
Stackable Banana Plug Connecting Wires

Red Stackable Banana Plug Connecting Wires, 36"

Price
$58.62
Log in for availability
Each (1UN)
Product Details & Documents

Flexible, Insulated Wire.

  • Insulated 4 mm banana plug at both ends
  • Plated spring contacts
  • Stack to accept other banana plugs
  • With side connectors
Specifications
  • Description:
    Red Stackable Banana Plug Connecting Wires
  • Length:
    36"