XXXX Quality Corks
11
Catalog # 470121-846
Supplier: MANTON INDUSTRIAL CORK SE
Supplier #: 55511
XXXX Quality Corks
Catalog # 470121-846
Supplier: MANTON INDUSTRIAL CORK SE
Jump to:
Product Details & Documents
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
- Fine Grain
- Ideal for Use as Tight-Fitting Stoppers
- Common Sizes
- Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
Specifications
- Stopper No.:11
- Cat. No.:470121-846
- Environmentally Preferable:Low Manufacturing Impact