Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ripple Tank
Ripple Tank
Ripple Tank, 40 x 40 cm
Catalog # 470121-500
Supplier:   VWR International
Ripple Tank
Catalog # 470121-500
Supplier:  VWR International
Ripple Tank
Ripple Tank
Ripple Tank, 40 x 40 cm
Price
$4,130.82
Log in for availability
Each (1UN)
Product Details & Documents
This compact tank can be used on an overhead projector or as a traditional ripple tank.
  • Large Viewing Area
  • For Use with or without an Overhead Projector
  • For All Wave-Phenomena Demonstrations
  • Ripple Motor
  • Halogen Lamp Unit
It features a ripple motor with an eccentric cam mounted on PVC bar along with three supports for the illuminator and the motor unit. Motor hanging clamps and springs for motor support ensure safe and secure use.
This controller provides independent, continuously variable DC outputs 0 to 5V for the ripple tank motor with a fixed AC output to operate a 6V, 20W bulb.
Ordering information: This kit includes a 6V halogen lamp unit, hand stroboscope, concave Perspex plate, convex Perspex plate, rectangular Perspex plate, curved reflector, large barrier, small barrier, plastic roller bar, illuminator lamp, and detachable 1.5 m mains lead. The ripple tank controller is required but not included.
Specifications
  • Description:
    Ripple Tank
  • Dimensions:
    40 x 40 cm