Order Entry
Mexico
Orders LinkContactUsLinkComponent
Resistance Blocks
Resistance Blocks
10 Ohm Resistance block
Catalog # 470121-092
Supplier:   VWR International
Resistance Blocks
Catalog # 470121-092
Supplier:  VWR International
Resistance Blocks
Resistance Blocks
10 Ohm Resistance block
Clearance
$2.00
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN)
Product Details & Documents
Economical yet Sturdy
  • Simple to use
  • Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Specifications
  • Description:
    10 Ohm Resistance block