Order Entry
Mexico
Orders LinkContactUsLinkComponent
Foam Brushes
Foam Brushes
Foam Cylinder Brush, Foam, Plastic
Catalog # 470120-528
Supplier:   BRUSHTECH
Supplier #:  B130C
Foam Brushes
Catalog # 470120-528
Supplier:  BRUSHTECH
Foam Brushes
Foam Brushes
Foam Cylinder Brush, Foam, Plastic
Price
$252.60
Log in for availability
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Foam brushes are designed for cleaning lab glass and plasticware.
  • Brushes will not chip, scratch or mar glassware
  • Handles are plastic, including grips
Brushes are made of a special foam that does not get wet, yet transports water and detergent to the glass surface. These brushes will easily out-live others. They should be a fixture in your prep room.
Specifications
  • Bristle Material:
    Foam
  • Handle Material:
    Plastic
  • Description:
    Foam Cylinder Brush